PRI444789553 Princeton BioMolecular Research, Inc.

4-Heptyl-benzene-1,3-diol

  • MDL number MFCD01684810
  • Cas Number : 18979-65-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart