PRI444789559 Princeton BioMolecular Research, Inc.

2-(2-Methylamino-ethyl)-isothiourea

  • MDL number MFCD19203978
  • Molecular Weight : 133.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart