PRI444789562 Princeton BioMolecular Research, Inc.

2-(2-Chloro-4-methoxy-phenoxy)-ethylamine

  • MDL number MFCD09906749
  • Molecular Weight : 201.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart