PRI444789576 Princeton BioMolecular Research, Inc.

N-(3,4-Dimethyl-phenyl)-2-hydroxy-benzamide

  • MDL number MFCD00124205
  • Cas Number : 282719-30-6
  • Molecular Weight : 241.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart