PRI444789597 Princeton BioMolecular Research, Inc.

2-(3-Amino-phenyl)-2,3-dihydro-benzo[e][1,3]oxazin-4-one

  • MDL number MFCD28248759
  • Cas Number : 408508-64-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart