PRI444789600 Princeton BioMolecular Research, Inc.

2-Chloro-N-[2-(4-chloro-phenyl)-2-oxo-ethyl]-acetamide

  • MDL number MFCD18432399
  • Molecular Weight : 246.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart