PRI444789610 Princeton BioMolecular Research, Inc.

3-[4-(2-Chloro-phenyl)-5-oxo-4,5-dihydro-tetrazol-1-yl]-propionic acid

  • MDL number MFCD28248772
  • Molecular Weight : 268.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart