PRI444789615 Princeton BioMolecular Research, Inc.

3-(3-Methyl-2,4-dioxo-3,4-dihydro-2H-quinazolin-1-yl)-propionic acid

  • MDL number MFCD28248778
  • Cas Number : 199915-83-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart