PRI444789627 Princeton BioMolecular Research, Inc.

2-Amino-6-hydroxymethyl-4-(4-methoxy-phenyl)-8-oxo-4,8-dihydro-pyrano[3,2-b]pyran-3-carboxylic acid ethyl ester

  • MDL number MFCD28248800
  • Molecular Weight : 373.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart