PRI444789631 Princeton BioMolecular Research, Inc.

3-[[3-aminopropoxy(dimethyl)silyl]oxy-dimethyl-silyl]oxypropan-1-amine

  • MDL number MFCD28248806
  • Molecular Weight : 280.51

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart