PRI444789634 Princeton BioMolecular Research, Inc.

5-[4-(1H-Benzoimidazol-2-yl)-butyl]-4-phenyl-4H-[1,2,4]triazole-3-thiol

  • MDL number MFCD28248810
  • Molecular Weight : 349.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart