PRI444789640 Princeton BioMolecular Research, Inc.

2-Isopropylamino-1-(5,6,7,8-tetrahydro-naphthalen-1-yl)-propan-1-ol

  • MDL number MFCD28248817
  • Molecular Weight : 247.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart