PRI444789641 Princeton BioMolecular Research, Inc.

(3-Oxo-3,4-dihydro-2H-benzo[1,4]oxazin-5-yloxy)-acetic acid methyl ester

  • MDL number MFCD28248818
  • Molecular Weight : 237.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart