PRI444789643 Princeton BioMolecular Research, Inc.

Benzyl-(2-thiocyanato-ethyl)-amine

  • MDL number MFCD28248821
  • Molecular Weight : 192.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart