PRI444789649 Princeton BioMolecular Research, Inc.

(6-Nitro-benzo[1,3]dioxol-5-yl)-acetic acid methyl ester

  • MDL number MFCD28248828
  • Cas Number : 40118-18-1
  • Molecular Weight : 239.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart