PRI638567975 Princeton BioMolecular Research, Inc.

1-Pyridin-4-ylmethyl-1,2,3,4-tetrahydro-isoquinoline

  • MDL number MFCD03541054
  • Molecular Weight : 224.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart