PRI638568053 Princeton BioMolecular Research, Inc.

4,5,6,7-tetrahydro-2H-indazol-3-amine

  • MDL number MFCD09880435
  • Cas Number : 41832-27-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart