PRI638568065 Princeton BioMolecular Research, Inc.

6-chloro-5-methylpyridine-3-sulfonyl chloride

  • MDL number MFCD09881190
  • Cas Number : 37105-10-5
  • Molecular Weight : 226.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart