PRI638568124 Princeton BioMolecular Research, Inc.

5-amino-2-methylisoxazol-3(2H)-one

  • MDL number MFCD12402091
  • Cas Number : 1216198-12-7
  • Molecular Weight : 114.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart