PRI638569020 Princeton BioMolecular Research, Inc.

3,7,11,15-Tetramethyl-hexadec-1-yn-3-ol

  • MDL number MFCD00129084
  • Cas Number : 29171-23-1
  • Molecular Weight : 294.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart