PRI638569196 Princeton BioMolecular Research, Inc.

[1-(4-Amino-furazan-3-yl)-5-phenyl-1H-[1,2,3]triazol-4-yl]-pyrrolidin-1-yl-methanone

  • MDL number MFCD01306058
  • Cas Number : 312275-05-1
  • Molecular Weight : 325.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart