PRI638569251 Princeton BioMolecular Research, Inc.

1-(4-Amino-furazan-3-yl)-5-phenyl-1H-[1,2,3]triazole-4-carboxylic acid hydrazide

  • MDL number MFCD00824524
  • Cas Number : 350475-45-5
  • Molecular Weight : 286.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart