PRI638569254 Princeton BioMolecular Research, Inc.

6-Methoxy-[1,2,5]oxadiazolo[3,4-b]pyrazin-5-ylamine

  • MDL number MFCD00461181
  • Cas Number : 339338-74-8
  • Molecular Weight : 167.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart