PRI638569263 Princeton BioMolecular Research, Inc.

1-(4-Amino-furazan-3-yl)-5-pyrrolidin-1-ylmethyl-1H-[1,2,3]triazole-4-carboxylic acid hydrazide

  • MDL number MFCD01450992
  • Cas Number : 309279-66-1
  • Molecular Weight : 293.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart