PRI638569296 Princeton BioMolecular Research, Inc.

3-(4-Ethoxy-phenyl)-6-propyl-cyclohex-2-enone

  • MDL number MFCD00429464
  • Cas Number : 177652-31-2
  • Molecular Weight : 258.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart