PRI638569409 Princeton BioMolecular Research, Inc.

3-(2-Benzo[1,3]dioxol-5-yl-1-cyano-vinyl)-benzonitrile

  • MDL number MFCD06629756
  • Cas Number : 833439-61-5
  • Molecular Weight : 274.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart