PRI638569546 Princeton BioMolecular Research, Inc.

3-(4-Isopropoxy-phenyl)-3-tetrazol-1-yl-propionic acid

  • MDL number MFCD04042431
  • Cas Number : 696649-41-9
  • Molecular Weight : 276.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart