PRI638569708 Princeton BioMolecular Research, Inc.

5-tert-Butyl-6a-hydroxy-3a-methyl-hexahydro-furo[2,3-c]pyrrol-4-one

  • MDL number MFCD00452208
  • Cas Number : 60999-30-6
  • Molecular Weight : 213.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart