PRI638569711 Princeton BioMolecular Research, Inc.

5-Ethyl-4-hydroxy-3-[2-(5-methoxy-1H-indol-3-yl)-ethyl]-4,5-dimethyl-oxazolidin-2-one

  • MDL number MFCD06618368
  • Cas Number : 883291-40-5
  • Molecular Weight : 332.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart