PRI638570015 Princeton BioMolecular Research, Inc.

4-Amino-benzoic acid 2,2,6,6-tetramethyl-piperidin-4-yl ester

  • MDL number MFCD08273447
  • Cas Number : 38879-01-5
  • Molecular Weight : 276.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart