PRI638570100 Princeton BioMolecular Research, Inc.

4-[5-(4-Fluoro-phenyl)-[1,2,4]oxadiazol-3-yl]-furazan-3-ylamine

  • MDL number MFCD02019210
  • Cas Number : 312628-68-5
  • Molecular Weight : 247.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart