PRI638570166 Princeton BioMolecular Research, Inc.

2-Amino-1-(3-chloro-phenyl)-4,5-dimethyl-1H-pyrrole-3-carbonitrile

  • MDL number MFCD08445345
  • Cas Number : 72578-54-2
  • Molecular Weight : 245.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart