PRI638570347 Princeton BioMolecular Research, Inc.

N-(Acetylamino-pyridin-2-yl-methyl)-acetamide

  • MDL number MFCD08848204
  • Cas Number : 924858-90-2
  • Molecular Weight : 207.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart