PRI638570366 Princeton BioMolecular Research, Inc.

4-Hydroxy-6-trifluoromethyl-pyridine-2-carboxylic acid amide

  • MDL number MFCD08447734
  • Cas Number : 924871-44-3
  • Molecular Weight : 206.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart