PRI638570538 Princeton BioMolecular Research, Inc.

1-[3-(1-Methoxy-ethyl)-[1,2,4]oxadiazol-5-yl]-ethylamine

  • MDL number MFCD21873068
  • Cas Number : 1528545-97-2
  • Molecular Weight : 171.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart