PRI638570641 Princeton BioMolecular Research, Inc.

2-(4-methyl-1-phenyl-pyrazol-3-yl)oxyacetic acid

  • MDL number MFCD28146186
  • Molecular Weight : 232.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart