PRI638570730 Princeton BioMolecular Research, Inc.

(17R)-17-hydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-3-one

  • MDL number MFCD00003662
  • Cas Number : 152-58-9
  • Molecular Weight : 346.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart