PRI638570742 Princeton BioMolecular Research, Inc.

(5R)-6,8-dioxabicyclo[3.2.1]octan-4-one

  • MDL number MFCD09028269
  • Cas Number : 53716-82-8
  • Molecular Weight : 128.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart