PRI638570764 Princeton BioMolecular Research, Inc.

(2S,5R)-2-(3,5-dimethyl-1H-pyrazol-1-yl)-6,8-dioxabicyclo[3.2.1]octan-4-one

  • MDL number MFCD08848258
  • Cas Number : 1046046-43-8
  • Molecular Weight : 222.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart