PRI638571223 Princeton BioMolecular Research, Inc.

5-Chloro-2-ethylsulfanyl-phenylamine

  • MDL number MFCD05259370
  • Cas Number : 1048664-15-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart