PRIH039FD4C1 Princeton BioMolecular Research, Inc.

2-Amino-N,N-dicyclohexyl-acetamide

  • MDL number MFCD13227219
  • Molecular Weight : 238.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart