PRIH039FD544 Princeton BioMolecular Research, Inc.

4-Hydroxy-1,6-dimethyl-2-oxo-1,2-dihydro-pyridine-3-carboxylic acid

  • MDL number MFCD06618410
  • Molecular Weight : 183.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart