PRIH039FD5FF Princeton BioMolecular Research, Inc.

1-methyl-1,5-dihydro-6H-[1,2]diazepino[4,5,6-cd]indol-6-one

  • MDL number MFCD21362922
  • Cas Number : 65923-22-0
  • Molecular Weight : 199.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart