PRIH039FD72A Princeton BioMolecular Research, Inc.

6-chloro-2-methyl-3-oxo-4H-1,4-benzoxazine-7-sulfonyl chloride

  • MDL number MFCD30728780
  • Molecular Weight : 296.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart