PRIH039FD745 Princeton BioMolecular Research, Inc.

tert-butyl 3-[[10-[[(3-tert-butoxy-3-oxo-propyl)amino]methyl]-9-anthryl]methylamino]propanoate

  • MDL number MFCD30745632
  • Molecular Weight : 492.66

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart