PRIH11AAF123 Princeton BioMolecular Research, Inc.

7-methylquinoline-2-carboxylic acid

  • MDL number MFCD09754110
  • Cas Number : 75434-10-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart