PRIH11AAF126 Princeton BioMolecular Research, Inc.

8-bromoquinoline-6-carboxylic acid

  • MDL number MFCD09754138
  • Cas Number : 791632-21-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart