PRIH11AAF130 Princeton BioMolecular Research, Inc.

3-hydroxy-4,7,7-trimethylbicyclo[2.2.1]heptane-2-carboxylic acid

  • MDL number MFCD01111358
  • Cas Number : 6246-41-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart