PRIH11AAF148 Princeton BioMolecular Research, Inc.

4-methylcinnoline

  • MDL number MFCD01736905
  • Cas Number : 14722-38-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart