PRIH11AAF168 Princeton BioMolecular Research, Inc.

(3-Methyl-1-phenyl-1H-pyrazol-4-yl)-acetic acid methyl ester

  • MDL number MFCD17267697
  • Cas Number : 1248551-08-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart